The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083532.1 | NP_001077001.1 | 8 - 9 | 1176 | 73 |
GenBank mRNA | BC123813.1 | - | 8 - 9 | 1176 | 73 |