Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAS2L1
Entrez Gene ID
518935
Gene Name
growth arrest specific 2 like 1
Gene Aliases
-
Chromosome Location
Chr.17: 68582956 - 68588420 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083698.1 NP_001077167.1 2 - 3 793 70
GenBank mRNA BC134474.1 - 2 - 3 793 70

View Full Product Details