Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HNRNPDL
Entrez Gene ID
534770
Gene Name
heterogeneous nuclear ribonucleoprotein D like
Gene Aliases
HNRPDL
Chromosome Location
Chr.6: 97209605 - 97217014 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083725.2 NP_001077194.1 5 - 6 1291 156
GenBank mRNA BC105386.1 - 5 - 6 770 156
BC134448.1 - 5 - 6 1292 156

View Full Product Details