Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GEM
Entrez Gene ID
538437
Gene Name
GTP binding protein overexpressed in skeletal muscle
Gene Aliases
-
Chromosome Location
Chr.14: 70066769 - 70080794 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083732.2 NP_001077201.2 4 - 5 812 68
NM_001276305.1 NP_001263234.1 4 - 5 738 68
GenBank mRNA BC134475.1 - 4 - 5 813 68
DV883786.1 - 4 - 5 684 68

View Full Product Details