Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC8D
Entrez Gene ID
539204
Gene Name
leucine rich repeat containing 8 VRAC subunit D
Gene Aliases
-
Chromosome Location
Chr.3: 53313187 - 53444422 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083741.1 NP_001077210.1 1 - 2 73 60
GenBank mRNA BC134543.1 - 1 - 2 73 60

View Full Product Details