Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFAND2A
Entrez Gene ID
613974
Gene Name
zinc finger AN1-type containing 2A
Gene Aliases
-
Chromosome Location
Chr.25: 41664244 - 41672227 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083762.1 NP_001077231.1 1 - 2 168 73
GenBank mRNA BC134789.1 - 1 - 2 168 73

View Full Product Details