Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNDIG1L
Entrez Gene ID
617660
Gene Name
synapse differentiation inducing 1 like
Gene Aliases
DSPC1, TMEM90A
Chromosome Location
Chr.10: 85710801 - 85768574 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083779.1 NP_001077248.1 2 - 3 624 70
GenBank mRNA BC134605.1 - 2 - 3 624 70

View Full Product Details