Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO1
Entrez Gene ID
788251
Gene Name
pleckstrin homology domain containing O1
Gene Aliases
-
Chromosome Location
Chr.3: 20475592 - 20485628 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083797.1 NP_001077266.1 4 - 5 520 63
GenBank mRNA BC134615.1 - 4 - 5 520 63

View Full Product Details