Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL1
Entrez Gene ID
282351
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 1
Gene Aliases
SIAT4A
Chromosome Location
Chr.14: 7864644 - 7955645 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097983.1 NP_001091452.1 1 - 2 319 91
GenBank mRNA AJ305086.1 - 1 - 3 263 91
BT030479.1 - 1 - 2 319 91

View Full Product Details