Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSMO1
Entrez Gene ID
504481
Gene Name
methylsterol monooxygenase 1
Gene Aliases
SC4MOL
Chromosome Location
Chr.17: 517366 - 534144 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098863.1 NP_001092333.1 1 - 2 125 114
GenBank mRNA BC146235.1 - 1 - 2 125 114

View Full Product Details