Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLBD1
Entrez Gene ID
317710
Gene Name
phospholipase B domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 95273631 - 95362983 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101044.2 NP_001094514.2 3 - 4 645 114
NM_001166298.1 NP_001159770.1 3 - 4 645 114
GenBank mRNA AJ299408.1 - 3 - 4 350 114
BC149435.1 - 3 - 4 631 114
EV650304.1 - 3 - 4 650 114

View Full Product Details