Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
F9
Entrez Gene ID
280688
Gene Name
coagulation factor IX
Gene Aliases
-
Chromosome Location
Chr.Unknown: 5524358 - 5558281 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103220.1 NP_001096690.1 4 - 5 404 70
GenBank mRNA BC149563.1 - 4 - 5 404 70
J00007.1 - 1 - 2 101 70

View Full Product Details