Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS42
Entrez Gene ID
504538
Gene Name
serine protease 42
Gene Aliases
PRSS42P, TESSP2
Chromosome Location
Chr.22: 52655482 - 52676224 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114626.2 NP_001108098.2 3 - 4 862 141

View Full Product Details