Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPPP
Entrez Gene ID
280968
Gene Name
tubulin polymerization promoting protein
Gene Aliases
p25
Chromosome Location
Chr.20: 71467587 - 71488493 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC133343.1 - 1 - 2 363 120

View Full Product Details