Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH5
Entrez Gene ID
281448
Gene Name
retinol dehydrogenase 5
Gene Aliases
9cRDH, P32
Chromosome Location
Chr.5: 57533652 - 57538050 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174166.3 NP_776591.2 1 - 2 86 70
GenBank mRNA EV670727.1 - 1 - 2 88 70

View Full Product Details