Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BFSP2
Entrez Gene ID
281645
Gene Name
beaded filament structural protein 2
Gene Aliases
LIFL-L
Chromosome Location
Chr.1: 135950794 - 136027701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174248.3 NP_776673.1 6 - 7 1254 128
GenBank mRNA BC122762.1 - 6 - 7 1254 128
X75160.1 - - 1301 128

View Full Product Details