Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAP1A
Entrez Gene ID
282031
Gene Name
RAP1A, member of RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 31588106 - 31676111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174448.5 NP_776873.1 7 - 8 737 78
GenBank mRNA BC134566.1 - 7 - 8 667 78
DR697686.1 - 3 - 4 279 78

View Full Product Details