Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCL
Entrez Gene ID
286773
Gene Name
phosducin like
Gene Aliases
-
Chromosome Location
Chr.11: 93986470 - 93995194 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174759.2 NP_777184.2 2 - 3 299 59
GenBank mRNA BC113227.1 - 2 - 3 299 59

View Full Product Details