Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGDS
Entrez Gene ID
286858
Gene Name
prostaglandin D2 synthase
Gene Aliases
-
Chromosome Location
Chr.11: 106021815 - 106024969 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174791.4 NP_777216.1 3 - 4 381 120
GenBank mRNA AB004647.1 - 3 - 4 362 120
BC133435.1 - 3 - 4 383 120
BC133438.1 - 3 - 4 383 120
BC133445.1 - 3 - 4 382 120
BC142054.1 - 3 - 4 382 120
BC142082.1 - 3 - 4 383 120
BC151436.1 - 3 - 4 382 120

View Full Product Details