Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAG8
Entrez Gene ID
337902
Gene Name
pregnancy-associated glycoprotein 8
Gene Aliases
-
Chromosome Location
Chr.29: 39726286 - 39736548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176619.3 NP_788792.2 4 - 5 498 136
GenBank mRNA AB098961.1 - 4 - 5 485 136
AF020510.2 - 4 - 5 454 136
BC133476.1 - 4 - 5 499 136

View Full Product Details