Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAG8
Entrez Gene ID
337902
Gene Name
pregnancy-associated glycoprotein 8
Gene Aliases
-
Chromosome Location
Chr.29: 39726286 - 39736548 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176619.3 NP_788792.2 5 - 6 694 97
GenBank mRNA AF020510.2 - 5 - 6 650 97
BC133476.1 - 5 - 6 695 97

View Full Product Details