Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAG10
Entrez Gene ID
337904
Gene Name
pregnancy-associated glycoprotein 10
Gene Aliases
-
Chromosome Location
Chr.29: 37551916 - 37560345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176621.3 NP_788794.2 3 - 4 367 64
GenBank mRNA AF020512.2 - 3 - 4 367 64

View Full Product Details