Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYVE1
Entrez Gene ID
404179
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
CRSBP-1, LYVE-1, XLKD1
Chromosome Location
Chr.15: 42102179 - 42116470 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205815.2 NP_991384.2 5 - 6 929 110
GenBank mRNA AY372937.1 - 5 - 6 817 110
BC140639.1 - 5 - 6 929 110

View Full Product Details