Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEPACAM
Entrez Gene ID
521015
Gene Name
hepatic and glial cell adhesion molecule
Gene Aliases
-
Chromosome Location
Chr.29: 28381633 - 28399064 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031759.2 NP_001026929.2 4 - 5 832 117
GenBank mRNA BC123427.1 - 4 - 5 832 117

View Full Product Details