Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3H1orf50
Entrez Gene ID
507125
Gene Name
chromosome 3 C1orf50 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 103464487 - 103472049 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034279.2 NP_001029451.2 2 - 3 199 136
GenBank mRNA BC102705.1 - 1 - 2 325 136
DT834744.1 - 2 - 3 201 136

View Full Product Details