Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
515913
Gene Name
thymus, brain and testes associated
Gene Aliases
C28H10orf27, SPATIAL
Chromosome Location
Chr.28: 26937387 - 26952734 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038121.2 NP_001033210.2 - 385 95
GenBank mRNA BC110031.1 - - 386 95

View Full Product Details