Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBATA
Entrez Gene ID
515913
Gene Name
thymus, brain and testes associated
Gene Aliases
C28H10orf27, SPATIAL
Chromosome Location
Chr.28: 26937387 - 26952734 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038121.2 NP_001033210.2 - 492 86
GenBank mRNA BC110031.1 - - 493 86
CK774174.1 - 6 - 7 754 86

View Full Product Details