Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTC4S
Entrez Gene ID
511749
Gene Name
leukotriene C4 synthase
Gene Aliases
-
Chromosome Location
Chr.7: 1519314 - 1522281 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046098.2 NP_001039563.2 2 - 3 231 62
GenBank mRNA BC111676.1 - 2 - 3 232 62
DV905069.1 - 2 - 3 174 62

View Full Product Details