Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PREB
Entrez Gene ID
525256
Gene Name
prolactin regulatory element binding
Gene Aliases
-
Chromosome Location
Chr.11: 72504550 - 72508647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076875.2 NP_001070343.2 4 - 5 859 98

View Full Product Details