Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA5B
Entrez Gene ID
514494
Gene Name
carbonic anhydrase 5B
Gene Aliases
-
Chromosome Location
Chr.X: 127724914 - 127752391 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080908.3 NP_001074377.2 3 - 4 471 142

View Full Product Details