Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALB
Entrez Gene ID
505238
Gene Name
RAS like proto-oncogene B
Gene Aliases
-
Chromosome Location
Chr.2: 71941008 - 72016193 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097985.1 NP_001091454.1 4 - 5 487 84
GenBank mRNA BC146173.1 - 3 - 4 406 84
BT030496.1 - 4 - 5 487 84

View Full Product Details