The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097988.1 | NP_001091457.1 | 7 - 8 | 1146 | 85 |
GenBank mRNA | BC140583.1 | - | 7 - 8 | 1146 | 85 |
BT030685.1 | - | 7 - 8 | 1252 | 85 |