Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMPO
Entrez Gene ID
510267
Gene Name
thymopoietin
Gene Aliases
LAP2beta
Chromosome Location
Chr.5: 62686655 - 62716051 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098005.1 NP_001091474.1 3 - 4 578 138
GenBank mRNA BC140628.1 - 3 - 4 578 138

View Full Product Details