Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHST14
Entrez Gene ID
511245
Gene Name
carbohydrate sulfotransferase 14
Gene Aliases
-
Chromosome Location
Chr.10: 36077444 - 36079402 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098012.1 NP_001091481.1 1 - 1 1751 134
GenBank mRNA BC133355.1 - 1 - 1 1751 134

View Full Product Details