Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX4
Entrez Gene ID
516030
Gene Name
distal-less homeobox 4
Gene Aliases
-
Chromosome Location
Chr.19: 36682225 - 36688507 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098041.1 NP_001091510.1 1 - 2 423 104
GenBank mRNA BC140682.1 - 1 - 2 423 104

View Full Product Details