Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COPS3
Entrez Gene ID
507932
Gene Name
COP9 signalosome subunit 3
Gene Aliases
-
Chromosome Location
Chr.19: 34886661 - 34904856 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098904.1 NP_001092374.1 7 - 8 819 69
GenBank mRNA BC146183.1 - 7 - 8 819 69

View Full Product Details