Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAH
Entrez Gene ID
508724
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
FAA
Chromosome Location
Chr.21: 26110597 - 26141035 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098912.1 NP_001092382.1 10 - 11 930 64
GenBank mRNA BC142487.1 - 10 - 11 930 64
BC151490.1 - 10 - 11 944 64
BC151824.1 - 10 - 11 930 64

View Full Product Details