Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM241B
Entrez Gene ID
510161
Gene Name
family with sequence similarity 241 member B
Gene Aliases
C28H10orf35
Chromosome Location
Chr.28: 25944553 - 25947529 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098923.1 NP_001092393.1 1 - 2 74 98
GenBank mRNA BC146161.1 - 1 - 2 74 98

View Full Product Details