Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
510283
Gene Name
tRNA methyltransferase 61A
Gene Aliases
GCD14, TRM61
Chromosome Location
Chr.21: 68166399 - 68172831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098927.2 NP_001092397.1 1 - 2 63 82
GenBank mRNA BC146157.1 - 1 - 2 65 82
BC151461.1 - 1 - 2 49 82

View Full Product Details