Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
510920
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK
Chromosome Location
Chr.8: 90058696 - 90067693 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098933.1 NP_001092403.1 3 - 4 637 71
GenBank mRNA BC146216.1 - 3 - 4 637 71

View Full Product Details