Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRLS1
Entrez Gene ID
512761
Gene Name
cardiolipin synthase 1
Gene Aliases
-
Chromosome Location
Chr.13: 48186986 - 48209893 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098951.1 NP_001092421.1 5 - 6 859 75
GenBank mRNA BC142224.1 - 5 - 6 859 75

View Full Product Details