Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH3
Entrez Gene ID
514771
Gene Name
G-patch domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 126314727 - 126321493 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098972.2 NP_001092442.1 5 - 6 1233 81
GenBank mRNA BC142442.1 - 5 - 6 1235 81

View Full Product Details