Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMD8
Entrez Gene ID
515469
Gene Name
FERM domain containing 8
Gene Aliases
-
Chromosome Location
Chr.29: 43582016 - 43603328 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098985.2 NP_001092455.1 1 - 2 125 72
GenBank mRNA BC142304.1 - 1 - 2 126 72

View Full Product Details