Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEK6
Entrez Gene ID
515816
Gene Name
NIMA related kinase 6
Gene Aliases
-
Chromosome Location
Chr.11: 95315110 - 95401331 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098988.2 NP_001092458.1 9 - 10 990 93
GenBank mRNA BC146269.1 - 9 - 10 992 93

View Full Product Details