Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TULP3
Entrez Gene ID
516380
Gene Name
TUB like protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 106845483 - 106876012 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098992.1 NP_001092462.1 5 - 6 629 114
GenBank mRNA BC142186.1 - 5 - 6 629 114

View Full Product Details