Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDSL
Entrez Gene ID
516783
Gene Name
serine dehydratase like
Gene Aliases
-
Chromosome Location
Chr.17: 61005840 - 61017877 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098995.1 NP_001092465.1 6 - 7 828 66
GenBank mRNA BC142489.1 - 6 - 7 828 66

View Full Product Details