Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBX1
Entrez Gene ID
517514
Gene Name
developing brain homeobox 1
Gene Aliases
-
Chromosome Location
Chr.29: 24757938 - 24762782 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098998.1 NP_001092468.1 2 - 3 765 86
GenBank mRNA BC142482.1 - 2 - 3 765 86

View Full Product Details