Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLX1
Entrez Gene ID
521806
Gene Name
distal-less homeobox 1
Gene Aliases
-
Chromosome Location
Chr.2: 24462466 - 24466469 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099017.1 NP_001092487.1 1 - 2 481 95
GenBank mRNA BC146096.1 - 1 - 2 481 95

View Full Product Details