Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL28
Entrez Gene ID
525011
Gene Name
kelch like family member 28
Gene Aliases
-
Chromosome Location
Chr.21: 54697362 - 54727376 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099030.1 NP_001092500.1 4 - 5 1612 85
GenBank mRNA BC146233.1 - 4 - 5 1612 85

View Full Product Details