Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCER1A
Entrez Gene ID
506783
Gene Name
Fc fragment of IgE receptor Ia
Gene Aliases
-
Chromosome Location
Chr.3: 10419630 - 10425913 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100310.2 NP_001093780.1 1 - 2 105 128
GenBank mRNA BC149946.1 - 1 - 2 106 128

View Full Product Details